accessibility menu, dialog, popup

UserName

CAS RN 507-52-8

CH3CH3HOFFF

CAS RN 507-52-8

IUPAC Name: 1,1,1-trifluoro-2-methylpropan-2-ol
Synonyms: 2-(trifluormethyl)-2-propanol
Molecular Weight (g/mol): 128.09
Molecular Formula: C4H7F3O
InChi Key: OCGWWLDZAFOHGD-UHFFFAOYSA-N
SMILES: CC(C)(O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results