missing translation for 'onlineSavingsMsg'
Learn More

2-Trifluoromethyl-2-propanol, 98%

Product Code. 15468929
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15468929

Brand: Thermo Scientific Alfa Aesar L10888.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 507-52-8 | C4H7F3O | 128.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 507-52-8
Density 1.17
Boiling Point 80°C to 81°C
Flash Point 15°C (59°F)
Odor Recognizable
Molecular Formula C4H7F3O
Refractive Index 1.34
MDL Number MFCD00014391
Quantity 5 g
UN Number UN1987
Beilstein 1736196
Synonym 2-trifluoromethyl-2-propanol, 2-trifluoromethylpropanol-2, 2-propanol, 1,1,1-trifluoro-2-methyl, 2-trifluormethyl-2-propanol, acmc-1aya6, 2,2,2-trifluoro-tert-butanol, dimethyl trifluoromethyl methanol, 2-trifluoromethyl propan-2-ol, 1-methyl-1-trifluoromethyl-ethanol
InChI Key OCGWWLDZAFOHGD-UHFFFAOYSA-N
SMILES CC(C)(O)C(F)(F)F
IUPAC Name 1,1,1-trifluoro-2-methylpropan-2-ol
Molecular Weight (g/mol) 128.09
PubChem CID 68182
Formula Weight 128.09
Percent Purity 98%
Chemical Name or Material 2-Trifluoromethyl-2-propanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.