accessibility menu, dialog, popup

UserName

CAS RN 507472-25-5

IUPAC Name: 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
Synonyms: 3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(2-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 432.43
Molecular Formula: C24H20N2O6
InChi Key: DRESTDPDCGPKNI-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results