accessibility menu, dialog, popup

UserName

H3CONOOOHO

CAS RN 5081-36-7

IUPAC Name: 3-methoxy-4-nitrobenzoic acid
Synonyms: 3-methoxy-4-nitrobenzoic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: PWURRRRGLCVBMX-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results