accessibility menu, dialog, popup

UserName

CAS RN 508203-48-3

CH3NFF

CAS RN 508203-48-3

IUPAC Name: 2,3-difluoro-4-methylbenzonitrile
Synonyms: 2,3-difluoro-4-methylbenzonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: WENILJFVUHUBBV-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results