CAS RN 50930-22-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-O-Methyl-Alpha-D-N-acetylneuraminic Acid, TRC
CAS: 50930-22-8 Molecular Formula: C12 H21 N O9 Molecular Weight (g/mol): 323.3 IUPAC Name: (2S,4S,5R,6R)-5-acetamido-4-hydroxy-2-methoxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid SMILES: CO[C@]1(C[C@H](O)[C@@H](NC(=O)C)[C@@H](O1)[C@H](O)[C@H](O)CO)C(=O)O