missing translation for 'onlineSavingsMsg'
Learn More

2-O-Methyl-Alpha-D-N-acetylneuraminic Acid, TRC

Product Code. 30479374
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479374 25 mg 25mg
30468544 10 mg 10mg
30477670 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479374 Supplier Toronto Research Chemicals Supplier No. TRCM27540025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50930-22-8
Molecular Formula C12 H21 N O9
Molecular Weight (g/mol) 323.3
IUPAC Name (2S,4S,5R,6R)-5-acetamido-4-hydroxy-2-methoxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CO[C@]1(C[C@H](O)[C@@H](NC(=O)C)[C@@H](O1)[C@H](O)[C@H](O)CO)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H21NO9/c1-5(15)13-8-6(16)3-12(21-2,11(19)20)22-10(8)9(18)7(17)4-14/h6-10,14,16-18H,3-4H2,1-2H3,(H,13,15)(H,19,20)/t6-,7+,8+,9+,10+,12-/m0/s1
Formula Weight 323.1216
Chemical Name or Material 2-O-Methyl-a-D-N-acetylneuraminic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.