accessibility menu, dialog, popup

UserName

NH2SHNNOO

CAS RN 51039-84-0

IUPAC Name: (2-nitrophenyl)thiourea
Synonyms: 2-nitrophenylthiourea
Molecular Weight (g/mol): 197.21
Molecular Formula: C7H7N3O2S
InChi Key: FLGZBEKWHFRZNP-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results