accessibility menu, dialog, popup

UserName

OOOOOOOOOOOOOHHOOHOHOHHOOHOHOHHOHOOHHOOHHOOHOHHO

CAS RN 51211-51-9

IUPAC Name: 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol hydrate
Synonyms: α-CD hydrate
Molecular Weight (g/mol): 990.86
Molecular Formula: C36H62O31
InChi Key: UQIQLAUJPPOBJR-UHFFFAOYNA-N
SMILES: O.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results