missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclodextrin hydrate, 98+%

Product Code. 10256380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10256380 25 g Glass bottle 25g
10246380 5 g Glass bottle 5g
10217023 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10256380 Supplier Thermo Scientific Acros Supplier No. 227290250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51211-51-9 | C36H60O30 | 972.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51211-51-9
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.85
MDL Number MFCD00150804
InChI Key HFHDHCJBZVLPGP-UHFFFAOYNA-N
Synonym alpha-cyclodextrin hydrate, water.beta-cyclodextrin, cyclohexaamylose hydrate, ^a-cyclodextrin hydrate, alpha-cyclodextrin water, 972.86 anhydrous
PubChem CID 60161466
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 278°C
Color White
Loss on Drying 11% max.
Infrared Spectrum Authentic
Assay Percent Range 0.5% max. Linear oligomers (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 14, 2718
Specific Rotation + 148
Solubility Information Solubility in water: 145g/L (25°C)
IUPAC Name 5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol hydrate
Formula Weight 972.85
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material α-Cyclodextrin hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.