accessibility menu, dialog, popup

UserName

CAS RN 51298-62-5

IUPAC Name: hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride
Synonyms: hydrogen methyl (2S)-2-amino-5-{[amino(nitroamino)methylidene]amino}pentanoate chloride
Molecular Weight (g/mol): 269.69
Molecular Formula: C7H16ClN5O4
InChi Key: QBNXAGZYLSRPJK-JEDNCBNOSA-N
SMILES: [H+].[Cl-].COC(=O)[C@@H](N)CCCN=C(N)N[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)