missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-NAME hydrochloride

Product Code. 30002178
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002178 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 30002178 Supplier Tocris Bioscience™ Supplier No. 0665/100
  • ONLINE EXCLUSIVE PRICE
    128.00 €
    120.75 € / 100mg
    Save 7.25 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Non-selective NOS inhibitor

NO synthase inhibitor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 51298-62-5
Quantity 100 mg
Molecular Formula C7H16ClN5O4
Formula Weight 269.69
Molecular Weight (g/mol) 269.69
Chemical Name or Material L-NAME hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.