accessibility menu, dialog, popup

UserName

CAS RN 513-35-9

IUPAC Name: 2-methylbut-2-ene
Synonyms: 2-methyl-2-butene
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N
SMILES: CC=C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (3)
  • (3)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (8)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)