missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene

Product Code. 11348297
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348297 500 g 500g
11338297 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348297 Supplier Thermo Scientific Alfa Aesar Supplier No. 031853.36
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 513-35-9 | C5H10 | 70.14 g/mol

2-Methyl-2-butene is used to prepare 3-Brom-2.3-dimethyl-1.1-dicyan-butan in the presence of 2,2'-azobis-(2,4-dimethyl-4-methoxyvaleronitrile) as a catalyst. It is used as a precursor for peroxyacetyl nitrate for the calibration purposes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C

Specifications

Melting Point -134°C
Color Colorless
Density 0.662 g/mL
Boiling Point 36°C to 40°C
Flash Point -45°C (-49°F)
Odor Mild
Refractive Index 1.387
Quantity 500 g
UN Number UN2460
Merck Index 14,607
Solubility Information Miscible with alcohol,benzene,ligroin,ethylene oxide and ether. Immiscible with water.
Formula Weight 70.13
Percent Purity 90%
Grade Technical
Physical Form Liquid
Assay Remainder mainly 2-methyl-1-butene
Chemical Name or Material 2-Methyl-2-butene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.