accessibility menu, dialog, popup

UserName

NH2H2NNOO

CAS RN 5131-58-8

IUPAC Name: 4-nitrobenzene-1,3-diamine
Synonyms: 4-nitro-1,3-benzenediamine
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: DPIZKMGPXNXSGL-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(N)=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results