missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-m-phenylenediamine, 95%

Product Code. 11318655
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318655 50 g 50g
11308655 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318655 Supplier Thermo Scientific Alfa Aesar Supplier No. L12166.18
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5131-58-8 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5131-58-8
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00025289
InChI Key DPIZKMGPXNXSGL-UHFFFAOYSA-N
Synonym 4-nitro-m-phenylenediamine, 4-nitro-1,3-phenylenediamine, 2,4-diaminonitrobenzene, 1,3-benzenediamine, 4-nitro, 4-nitro-1,3-benzenediamine, 1,3-diamino-4-nitrobenzene, m-phenylenediamine, 4-nitro, 3-amino-4-nitrophenylamine, 4-nitro-1,3-fenylendiamin, unii-0d5u5ew62z
PubChem CID 21208
IUPAC Name 4-nitrobenzene-1,3-diamine
SMILES NC1=CC=C(C(N)=C1)[N+]([O-])=O

Specifications

Melting Point 156°C to 160°C
Quantity 50 g
Beilstein 2094904
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 4-Nitro-m-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.