CAS RN 51436-99-8
IUPAC Name:
4-bromo-2-fluoro-1-methylbenzene
Synonyms:
toluene,2-fluoro-4-bromo
Molecular Weight (g/mol):
189.03
Molecular Formula:
C7H6BrF
InChi Key:
YZFVUQSAJMLFOZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Br)C=C1F
1
–
1
of
1
results
1
4-Bromo-2-fluorotoluene, 99%
CAS: 51436-99-8 Molecular Formula: C7H6BrF Molecular Weight (g/mol): 189.027 MDL Number: MFCD00013551 InChI Key: YZFVUQSAJMLFOZ-UHFFFAOYSA-N Synonym: 4-bromo-2-fluorotoluene,2-fluoro-4-bromotoluene,benzene, 4-bromo-2-fluoro-1-methyl,toluene,2-fluoro-4-bromo,4-bromo-2-fluoro-1-methyl-benzene,pubchem1601,acmc-1an2v,2-fluoro-4-bromo toluene,4-bromo-2-fluoro-toluene PubChem CID: 171040 IUPAC Name: 4-bromo-2-fluoro-1-methylbenzene SMILES: CC1=C(C=C(C=C1)Br)F