accessibility menu, dialog, popup

UserName

ONNHNN

CAS RN 51449-77-5

IUPAC Name: 1-phenyl-2,5-dihydro-1H-1,2,3,4-tetrazol-5-one
Synonyms: 1-phenyl-2H-1,2,3,4-tetrazol-5-one
Molecular Weight (g/mol): 162.15
Molecular Formula: C7H6N4O
InChi Key: FYYACQQSYSDVJB-UHFFFAOYSA-N
SMILES: O=C1N=NNN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results