missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(1H-Tetrazol-5-yl)phenol, 97%

Product Code. 15453747
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453747 250 mg 250mg
11352749 1 g 1g
11362749 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15453747 Supplier Alfa Aesar™ Supplier No. H27672.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51449-77-5
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.152
MDL Number MFCD09037787
InChI Key SQPPXOJPOBGAAU-UHFFFAOYSA-N
Synonym phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol
PubChem CID 42553059
IUPAC Name 6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C2NNN=N2)C(=O)C=C1

Specifications

Melting Point ∽230°C (decomposition)
Quantity 250 mg
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 2-(1H-Tetrazol-5-yl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.