accessibility menu, dialog, popup

UserName

H3CHOOSCl

CAS RN 51527-19-6

IUPAC Name: 2-(5-chloro-3-methyl-1-benzothiophen-2-yl)acetic acid
Synonyms: tianafac
Molecular Weight (g/mol): 240.7
Molecular Formula: C11H9ClO2S
InChi Key: QNJIHQOPIPJYLU-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)SC2=CC=C(Cl)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results