missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid, 95%, Thermo Scientific™

Product Code. 10643413
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10643413 1 g Amber Glass Bottle 1g
10324392 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10643413 Supplier Thermo Scientific Maybridge Supplier No. MWP00226DA
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51527-19-6
Molecular Formula C11H9ClO2S
Molecular Weight (g/mol) 240.70
MDL Number MFCD00052300
InChI Key QNJIHQOPIPJYLU-UHFFFAOYSA-N
Synonym 2-5-chloro-3-methylbenzo b thiophen-2-yl acetic acid, tianafac, 5-chloro-3-methylbenzo b thiophene-2-acetic acid, unii-p0t3zrk3xv, p0t3zrk3xv, 2-5-chloro-3-methyl-1-benzothiophen-2-yl acetic acid, tianafacum, tianafaco, tianafac inn
PubChem CID 68686
IUPAC Name 2-(5-chloro-3-methyl-1-benzothiophen-2-yl)acetic acid
SMILES CC1=C(CC(O)=O)SC2=CC=C(Cl)C=C12

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 240.71
Percent Purity 95%
Chemical Name or Material 2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.