accessibility menu, dialog, popup

UserName

OO(Z)

CAS RN 51557-26-7

IUPAC Name: (2Z)-3-(naphthalen-2-yl)prop-2-enoate
Synonyms: (2Z)-3-(naphthalen-2-yl)prop-2-enoate
Molecular Weight (g/mol): 197.21
Molecular Formula: C13H9O2
InChi Key: KWGPBDBAAXYWOJ-VURMDHGXSA-M
SMILES: [O-]C(=O)\C=C/C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results