accessibility menu, dialog, popup

UserName

CAS RN 5162-82-3

CH3ClOO

CAS RN 5162-82-3

IUPAC Name: 3-chloro-4-methylbenzoate
Synonyms: 3-chloro-4-methylbenzoate
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H6ClO2
InChi Key: SDKUOEOJAXGCLU-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1Cl)C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results