CAS RN 5164-76-1
CAS RN 5164-76-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Dimethyl Glutaconate (~10% Cis), TRC
CAS: 5164-76-1 Molecular Formula: C7H10O4 Molecular Weight (g/mol): 158.15 Synonym: NSC 30046,Dimethyl 2-Pentene-1,5-dioate,Dimethyl 2-Pentenedioate,Glutaconic Acid Dimethyl Ester,2-Pentenedioic Acid Dimethyl Ester,2-Pentenedioic Acid 1,5-Dimethyl Ester SMILES: COC(=O)C\C=C\C(=O)OC
2
Dimethyl Glutaconate (~25% cis), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dimethyl Glutaconate (~25% cis), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.