accessibility menu, dialog, popup

UserName

CAS RN 517-51-1

IUPAC Name: 5,6,11,12-tetraphenyltetracene
Synonyms: rubrene
Molecular Weight (g/mol): 532.69
Molecular Formula: C42H28
InChi Key: YYMBJDOZVAITBP-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C(C3=CC=CC=C3)=C3C=CC=CC3=C(C3=CC=CC=C3)C2=C(C2=CC=CC=C2)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)