missing translation for 'onlineSavingsMsg'
Learn More

Rubrene, 97%

Product Code. 15430408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430408 25 g 25g
15410408 1 g 1g
15420408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430408 Supplier Thermo Scientific Alfa Aesar Supplier No. H53507.14
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517-51-1 | C42H28 | 532.686 g/mol

Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rubrene is an organic semiconductor used in organic light-emitting diodes (OLEDs) and organic field-effect transistors. It is used as a sensitizer in chemoluminesence. It is also used to prepare single crystal transistors. It acts as a reagent for chemiluminescence research and for transition metal complex ligation. Further, it is used as a p-type organic semiconductor.

Solubility
Soluble in hot toluene. Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-51-1
Molecular Formula C42H28
Molecular Weight (g/mol) 532.686
MDL Number MFCD00003703
InChI Key YYMBJDOZVAITBP-UHFFFAOYSA-N
Synonym rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
PubChem CID 68203
IUPAC Name 5,6,11,12-tetraphenyltetracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8

Specifications

Melting Point 330°C to 335°C
Quantity 25 g
Beilstein 1917339
Sensitivity Air Sensitive
Solubility Information Soluble in hot toluene. Insoluble in water.
Formula Weight 532.68
Percent Purity 97%
Chemical Name or Material Rubrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.