accessibility menu, dialog, popup

UserName

CAS RN 51781-21-6

HClH3CCH3H3CNHOHOHNO

CAS RN 51781-21-6

IUPAC Name: hydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
Synonyms: hydrogen carteolol chloride
Molecular Weight (g/mol): 328.84
Molecular Formula: C16H25ClN2O3
InChi Key: FYBXRCFPOTXTJF-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)