accessibility menu, dialog, popup

UserName

OOHO

CAS RN 51939-71-0

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-2-carboxylic acid
Synonyms: chroman-2-carboxylic acid
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: SFLFCQJQOIZMHF-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)