missing translation for 'onlineSavingsMsg'
Learn More

Chromane-2-carboxylic Acid, TRC

Product Code. 30367778
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367778 500 mg 500mg
30349724 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367778 Supplier Toronto Research Chemicals Supplier No. TRCC427970500MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chromane-2-carboxylic Acid
CAS 51939-71-0
Molecular Formula C10 H10 O3
Quantity 500 mg
Synonym 2H-1-Benzopyran-2-carboxylic acid, 3,4-dihydro-, 2-Chromancarboxylic acid (6CI), 3,4-Dihydro-2H-1-benzopyran-2-carboxylic acid, (±)-2-Chromancarboxylic acid, 3,4-Dihydro-1-benzopyran-2-carboxylic acid, Chromane-2-carboxylic acid
InChI Formula InChI=1S/C10H10O3/c11-10(12)9-6-5-7-3-1-2-4-8(7)13-9/h1-4,9H,5-6H2,(H,11,12)
SMILES OC(=O)C1CCc2ccccc2O1
IUPAC Name 3,4-dihydro-2H-chromene-2-carboxylic acid
Molecular Weight (g/mol) 178.18
Formula Weight 178.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.