accessibility menu, dialog, popup

UserName

CAS RN 5202-89-1

CH3OOClH2N

CAS RN 5202-89-1

IUPAC Name: methyl 2-amino-5-chlorobenzoate
Synonyms: methyl 2-amino-5-chlorobenzoate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: IGHVUURTQGBABT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Cl)=CC=C1N
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)