accessibility menu, dialog, popup

UserName

H3CON

CAS RN 5203-14-5

IUPAC Name: 4-butoxybenzonitrile
Synonyms: 4-butoxybenzonitrile
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: RRGQINKVTNAIBB-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results