accessibility menu, dialog, popup

UserName

HClH2NNHO

CAS RN 52068-30-1

IUPAC Name: hydrogen [4-(benzyloxy)phenyl]hydrazine chloride
Synonyms: hydrogen [4-(benzyloxy)phenyl]hydrazine chloride
Molecular Weight (g/mol): 250.73
Molecular Formula: C13H15ClN2O
InChi Key: OVNUPJXMCMTQCN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(OCC2=CC=CC=C2)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results