accessibility menu, dialog, popup

UserName

CH3OON(E/Z)

CAS RN 52116-81-1

IUPAC Name: methyl 3-(3-cyanophenyl)prop-2-enoate
Synonyms: methyl 3-(3-cyanophenyl)prop-2-enoate
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: BQJYULBYGRTLPZ-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=CC(=C1)C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results