missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyanocinnamate, Thermo Scientific™

Product Code. 11429560
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429560 1 g 1g
11419560 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429560 Supplier Thermo Scientific Alfa Aesar Supplier No. H50872.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52116-81-1
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD08064268
InChI Key BQJYULBYGRTLPZ-UHFFFAOYSA-N
Synonym methyl 3-3-cyanophenyl acrylate, 3-3-cyano-phenyl-acrylic acid methyl ester, methyl 3-cyanocinnamate, e-methyl 3-3-cyanophenyl acrylate, methyl 2e-3-3-cyanophenyl prop-2-enoate, 3-cyanocinnamic acid methyl ester, methyl e-3-3-cyanophenyl acrylate, methyl 2e-3-3-cyanophenyl acrylate, 3-3-cyanophenyl acrylic acid methyl ester
PubChem CID 13415097
IUPAC Name methyl (E)-3-(3-cyanophenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC=CC(=C1)C#N

Specifications

Quantity 1 g
Formula Weight 187.2
Chemical Name or Material Methyl 3-cyanocinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.