CAS RN 52221-92-8
IUPAC Name:
3-(2-bromophenyl)propan-1-ol
Synonyms:
3-(2-bromophenyl)propan-1-ol
Molecular Weight (g/mol):
215.09
Molecular Formula:
C9H11BrO
InChi Key:
VODAJGPTULSNSU-UHFFFAOYSA-N
SMILES:
OCCCC1=CC=CC=C1Br
1
–
1
of
1
results
1
3-(2-Bromophenyl)-1-propanol, 98%, Thermo Scientific™
CAS: 52221-92-8 Molecular Formula: C9H11BrO Molecular Weight (g/mol): 215.09 MDL Number: MFCD09028685 InChI Key: VODAJGPTULSNSU-UHFFFAOYSA-N Synonym: 3-2-bromo-phenyl-propan-1-ol,3-2-bromophenyl propan-1-ol,3-2-bromophenyl-1-propanol,benzenepropanol, 2-bromo,ksc268e5r,3-2-bromo-phenyl ;-propan-1-ol PubChem CID: 10512846 IUPAC Name: 3-(2-bromophenyl)propan-1-ol SMILES: C1=CC=C(C(=C1)CCCO)Br