accessibility menu, dialog, popup

UserName

CAS RN 52222-73-8

IUPAC Name: 4-(trifluoromethyl)-1H-pyrazole
Synonyms: 4-(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol): 136.08
Molecular Formula: C4H3F3N2
InChi Key: KDEJQUNODYXYBJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNN=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results