missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-1H-pyrazole, 97%

Product Code. 15417337
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417337 1 g 1g
15427337 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417337 Supplier Thermo Scientific Alfa Aesar Supplier No. H34396.03
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52222-73-8 | C4H3F3N2 | 136.077 g/mol

It is used as pharmaceutical intermediates. The palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazole with aryl iodides provided high yields of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. The palladium-catalyzed cross-coupling reactions of 5-tributylstannyl-4-fluoro-1H-pyrazole with aryl iodides provided high yields of the corresponding 5-aryl-4-fluoro-1H-pyrazoles.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52222-73-8
Molecular Formula C4H3F3N2
Molecular Weight (g/mol) 136.077
MDL Number MFCD11226572
InChI Key KDEJQUNODYXYBJ-UHFFFAOYSA-N
Synonym 4-trifluoromethyl-1h-pyrazole, 1h-pyrazole, 4-trifluoromethyl, 4-trifluoromethylpyrazole, 4-trifluoromethyl pyrazole, 4-trifluoromethyl-1 h-pyrazole, 4-trifluoromethyl ;-1h-pyrazole
PubChem CID 12777795
IUPAC Name 4-(trifluoromethyl)-1H-pyrazole
SMILES C1=C(C=NN1)C(F)(F)F

Specifica

Melting Point 76°C to 78°C
Quantity 1 g
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 4-Trifluoromethyl-1H-pyrazole

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.