accessibility menu, dialog, popup

UserName

H3COHN

CAS RN 5223-06-3

IUPAC Name: 2-(5-ethylpyridin-2-yl)ethan-1-ol
Synonyms: 2-(5-ethylpyridin-2-yl)ethanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: OUJMXIPHUCDRAS-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CCO)N=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)