accessibility menu, dialog, popup

UserName

H3COOH3C

CAS RN 52289-54-0

IUPAC Name: 4-methoxy-2-methylbenzaldehyde
Synonyms: 4-methoxy-2-methylbenzaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: WICYVKGMEJSDAO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(C)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)