accessibility menu, dialog, popup

UserName

BrBrNONBrBrONBrBrO

CAS RN 52434-90-9

IUPAC Name: tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
Synonyms: tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 728.69
Molecular Formula: C12H15Br6N3O3
InChi Key: NZUPFZNVGSWLQC-UHFFFAOYNA-N
SMILES: BrCC(Br)CN1C(=O)N(CC(Br)CBr)C(=O)N(CC(Br)CBr)C1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results