missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(2,3-dibromopropyl) Isocyanurate, TRC

Product Code. 30452217
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452217 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452217 Supplier Toronto Research Chemicals Supplier No. TRCT79895010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris(2,3-dibromopropyl) isocyanurate
CAS 52434-90-9
Molecular Formula C12 H15 Br6 N3 O3
Quantity 10 g
Synonym 1,3,5-Tris(2,3-dibromopropyl)-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 1,3,5-Tris(2,3-dibromopropyl) isocyanurate, 1,3,5-Tris(2,3-dibromopropyl)-1,3,5-triazine-2,4,6-trione, 1,3,5-Tris(2,3-dibromopropyl)-2,4,6-trioxohexahydro-s-triazine, 1,3,5-Tris(2,3-dibromopropyl)-2,4,6-trioxohexahydrotriazine, AFR 1002, EB 70, FC 140R, FCP 660, FCP 660CN, FR 930, Fire Cut P 660, Fire Cut P 660CN, M 2937, Pyroguard SR 750, TAIC 6B, TBC, TBC (fireproofing agent), Tri(2,3-dibromopropyl) isocyanurate, Tris(2,3-dibromopropyl) isocyanurate
InChI Formula InChI=1S/C12H15Br6N3O3/c13-1-7(16)4-19-10(22)20(5-8(17)2-14)12(24)21(11(19)23)6-9(18)3-15/h7-9H,1-6H2
SMILES BrCC(Br)CN1C(=O)N(CC(Br)CBr)C(=O)N(CC(Br)CBr)C1=O
IUPAC Name 1,3,5-tris(2,3-dibromopropyl)-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol) 728.69
Formula Weight 722.6214
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.