accessibility menu, dialog, popup

UserName

ONONH2

CAS RN 52481-41-1

IUPAC Name: 4-[2-(morpholin-4-yl)ethoxy]aniline
Synonyms: 4-[2-(morpholin-4-yl)ethoxy]aniline
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: ZHFFNLQQANCJEQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(OCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results