missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Morpholin-4-ylethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10669744
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10669744 250 mg 250mg
10193954 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10669744 Supplier Thermo Scientific Maybridge Supplier No. CC42114CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 52481-41-1
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.288
InChI Key ZHFFNLQQANCJEQ-UHFFFAOYSA-N
Synonym 4-2-morpholinoethoxy aniline, 4-2-morpholin-4-ylethoxy aniline, 4-2-morpholin-4-yl-ethoxy-phenylamine, 4-2-morpholin-4-yl ethoxy aniline, 4-2-4-morpholinyl ethoxy aniline, 4-2-morpholin-4-yl-ethoxy phenylamine, 4-2-morpholin-4-ylethoxy phenylamine, 4-2-4-amino-phenoxy ethyl-morpholine
PubChem CID 6484711
IUPAC Name 4-(2-morpholin-4-ylethoxy)aniline
SMILES C1COCCN1CCOC2=CC=C(C=C2)N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material 4-(2-Morpholin-4-ylethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.