accessibility menu, dialog, popup

UserName

CAS RN 526-78-3

HOOBrBrOHO

CAS RN 526-78-3

IUPAC Name: 2,3-dibromobutanedioic acid
Synonyms: 2,3-dibromosuccinic acid
Molecular Weight (g/mol): 275.88
Molecular Formula: C4H4Br2O4
InChi Key: FJWGRXKOBIVTFA-UHFFFAOYNA-N
SMILES: OC(=O)C(Br)C(Br)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results