accessibility menu, dialog, popup

UserName

NH2ClNN

CAS RN 5305-59-9

IUPAC Name: 6-chloropyrimidin-4-amine
Synonyms: 6-chloropyrimidin-4-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: DUKKRSPKJMHASP-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)