missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloropyrimidine, 98%, Thermo Scientific™

Product Code. 11420010
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420010 1 g 1g
11410010 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420010 Supplier Thermo Scientific Alfa Aesar Supplier No. H50551.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5305-59-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00053576
InChI Key DUKKRSPKJMHASP-UHFFFAOYSA-N
Synonym 4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630
PubChem CID 238012
IUPAC Name 6-chloropyrimidin-4-amine
SMILES C1=C(N=CN=C1Cl)N

Specifikationer

Quantity 1 g
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 4-Amino-6-chloropyrimidine
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.