accessibility menu, dialog, popup

UserName

CH3ClHO

CAS RN 5306-98-9

IUPAC Name: 5-chloro-2-methylphenol
Synonyms: 5-chloro-2-methylphenol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: KKFPXGXMSBBNJI-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1O
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)