accessibility menu, dialog, popup

UserName

H3COOSNH2N

CAS RN 53266-94-7

IUPAC Name: ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
Synonyms: ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: SHQNGLYXRFCPGZ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CSC(N)=N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (6)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)