missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-4-thiazoleacetate, TRC

Product Code. 30375018
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375018 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375018 Supplier Toronto Research Chemicals Supplier No. TRCE8993302.5G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-Amino-4-thiazoleacetate
CAS 53266-94-7
Molecular Formula C7H10N2O2S
Quantity 2.5 g
Synonym 2-Amino-4-thiazoleacetic Acid Ethyl Ester, (2-Amino-4-thiazolyl)acetic Acid Ethyl ester, (2-Amino-thiazole-4-yl)acetic Acid Ethyl Ester, 2-(2-Aminothiazol-4-yl)acetic Acid Ethyl Ester, 2-Amino-4-(carbethoxymethyl)thiazole, 2-Amino-4-(ethoxycarbonylmethyl)-1,3-thiazole, 2-Amino-4-[(ethoxycarbonyl)methyl]thiazole, 2-Amino-4-thiazoleacetic Acid Ethyl Ester, Ethyl (2-amino-1,3-thiazol-4-yl)acetate
InChI Formula InChI=1S/C7H10N2O2S/c1-2-11-6(10)3-5-4-12-7(8)9-5/h4H,2-3H2,1H3,(H2,8,9)
SMILES CCOC(=O)Cc1csc(N)n1
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol) 186.23
Formula Weight 186.0463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.