accessibility menu, dialog, popup

UserName

CAS RN 53312-82-6

NH2BrN

CAS RN 53312-82-6

IUPAC Name: 4-amino-2-bromobenzonitrile
Synonyms: 4-amino-2-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: OSYGPHDNNWRFJO-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results